5-(Benzyloxy)-6-bromo-2-pyridinecarbaldehyde ≥95%

Supplier: Matrix Scientific

048648-500MG 048648-1G
101884-722EA 921.54 USD
101884-722 101884-724
5-(Benzyloxy)-6-bromo-2-pyridinecarbaldehyde ≥95%
5-(Benzyloxy)-6-bromo-2-pyridinecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀BrNO₂
Melting Pt: 109…111 °C
MDL Number: MFCD13193072
CAS Number: 1192263-80-1

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR