(2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-yl)(phenyl)methanone ≥97%
Supplier: Matrix Scientific
048647-500MG
048647-1G
101884-718EA
375.38
USD
101884-718
101884-720
(2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-yl)(phenyl)methanone ≥97%
(2-Amino-4,7-dihydro-5H-thieno[2,3-c]pyran-3-yl)(phenyl)methanone
Hazard: Irritant
Formula:
C₁₄H₁₃NO₂S Melting Pt: 172…173 °C |
MDL Number:
MFCD13193071 CAS Number: 893387-73-0 |
Specification Test Results
Melting point | 172-173°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...