[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-yl)methanol

048642-500MG 048642-1G 048642-5G
101884-702EA 264.62 USD
101884-702 101884-704 101884-706
[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol ≥95%
[4'-(Trifluoromethyl)[1,1'-biphenyl]-4-yl]methanol

Hazard: Irritant

Formula: C₁₄H₁₁F₃O
MW: 252.24 g/mol
Melting Pt: 147…149 °C
MDL Number: MFCD01862515
CAS Number: 457889-46-2

Order Now

Specification Test Results

Melting point 147-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR