4-Nitro-2-(trichloroacetyl)pyrrole ≥95%
Supplier: Matrix Scientific
Synonyms:
2,2,2-Trichloro-1-(4-nitro-1H-pyrrol-2-yl)ethanone
048641-500MG
048641-1G
048641-5G
101884-696EA
181.54
USD
101884-696
101884-698
101884-700
4-Nitro-2-(trichloroacetyl)pyrrole ≥95%
4-Nitro-2-(trichloroacetyl)pyrrole
Hazard: Irritant
Formula:
C₆H₃Cl₃N₂O₃ Melting Pt: 178-179°C |
MDL Number:
MFCD05663491 CAS Number: 53391-50-7 |
Specification Test Results
Melting point | 178-179°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...