tert-Butyl-N-[4-cyano-2-iodo-5-(trifluoromethyl)phenyl]carbamate ≥95%
Supplier: Matrix Scientific
048639-500MG
048639-1G
101884-692EA
747.69
USD
101884-692
101884-694
tert-Butyl-N-[4-cyano-2-iodo-5-(trifluoromethyl)phenyl]carbamate ≥95%
tert-Butyl-N-[4-cyano-2-iodo-5-(trifluoromethyl)phenyl]carbamate
Hazard: Irritant
Formula:
C₁₃H₁₂F₃IN₂O₂ MW: 412.15 g/mol Melting Pt: 154…155 °C |
MDL Number:
MFCD12922694 CAS Number: 1190198-36-7 |
Specification Test Results
Melting point | 154-155°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...