2-Sulfanyl-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Mercapto-6,7-dihydro-3H-cyclopentapyrimidin-4(5H)-one

048638-500MG 048638-1G
101884-688EA 375.38 USD
101884-688 101884-690
2-Sulfanyl-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-ol ≥95%
2-Sulfanyl-6,7-dihydro-5H-cyclopenta[d]pyrimidin-4-ol

Hazard: Irritant

Formula: C₇H₈N₂OS
MW: 168.22 g/mol
Melting Pt: >320 °C
MDL Number: MFCD00223637
CAS Number: 35563-27-0

Order Now

Specification Test Results

Melting point >320°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR