4-Piperazino-2-(trifluoromethyl)quinazoline ≥95%

Supplier: Matrix Scientific

048637-500MG 048637-1G
101884-684EA 1109.23 USD
101884-684 101884-686
4-Piperazino-2-(trifluoromethyl)quinazoline ≥95%
4-Piperazino-2-(trifluoromethyl)quinazoline

Hazard: Irritant

Formula: C₁₃H₁₃F₃N₄
Melting Pt: 105…107 °C
MDL Number: MFCD06446619
CAS Number: 401567-99-5

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR