4-Nitropyrrole-2-carboxylic acid hydrate ≥95%
Supplier: Matrix Scientific
048636-500MG
048636-1G
048636-5G
101884-678EA
353.85
USD
101884-678
101884-680
101884-682
4-Nitropyrrole-2-carboxylic acid hydrate ≥95%
4-Nitropyrrole-2-carboxylic acid hydrate
Hazard: Irritant
Formula:
C₅H₄N₂O₄ MW: 156.10 g/mol Melting Pt: 217…218 °C |
MDL Number:
MFCD00059928 CAS Number: 5930-93-8 |
Specification Test Results
Melting point | 217-218°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...