4-Nitro-1H-pyrrole-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
048635-500MG
048635-1G
101884-674EA
556.92
USD
101884-674
101884-676
4-Nitro-1H-pyrrole-2-carbohydrazide ≥95%
4-Nitro-1H-pyrrole-2-carbohydrazide
Hazard: Irritant
Formula:
C₅H₆N₄O₃ Melting Pt: 286…288 °C |
MDL Number:
MFCD07339177 CAS Number: 28494-48-6 |
Specification Test Results
Melting point | 286-288°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...