Methyl-4-nitro-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-nitro-1H-pyrrole-2-carboxylate

048633-500MG 048633-1G
101884-668EA 547.69 USD
101884-668 101884-670
Methyl-4-nitro-1H-pyrrole-2-carboxylate ≥95%
Methyl-4-nitro-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₆H₆N₂O₄
MW: 170.13 g/mol
Melting Pt: 192…193 °C
MDL Number: MFCD08753843
CAS Number: 13138-74-4

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR