Methyl-1-methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate ≥95%
Supplier: Matrix Scientific
048632-500MG
048632-1G
101884-664EA
747.69
USD
101884-664
101884-666
Methyl-1-methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate ≥95%
Methyl-1-methyl-2-oxo-2,3-dihydro-1H-1,3-benzimidazole-5-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₃ Melting Pt: 253…255 °C |
MDL Number:
MFCD12922711 CAS Number: 396652-38-3 |
Specification Test Results
Melting point | 253-255°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...