2-(Benzyloxy)-4-methoxybenzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Benzyloxy)-4-methoxybenzaldehyde
048631-500MG
048631-1G
048631-5G
101884-658EA
272.31
USD
101884-658
101884-660
101884-662
2-(Benzyloxy)-4-methoxybenzenecarbaldehyde ≥95%
2-(Benzyloxy)-4-methoxybenzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₅H₁₄O₃ MW: 242.28 g/mol Melting Pt: 69…71 °C |
MDL Number:
MFCD02113682 CAS Number: 32884-23-4 |
Specification Test Results
Melting point | 69-71°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...