Methyl-4,6-dihydroxy-5-nitronicotinate ≥95%
Supplier: Matrix Scientific
048629-500MG
048629-1G
048629-5G
101884-648EA
278.46
USD
101884-648
101884-650
101884-652
Methyl-4,6-dihydroxy-5-nitronicotinate ≥95%
Methyl-4,6-dihydroxy-5-nitronicotinate
Hazard: Irritant
Formula:
C₇H₆N₂O₆ MW: 214.14 g/mol Melting Pt: 289…292 °C |
MDL Number:
MFCD12922702 CAS Number: 89247-04-1 |
Specification Test Results
Melting point | 289-292°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...