Methyl-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate ≥95%
Supplier: Matrix Scientific
048627-500MG
048627-1G
101884-640EA
649.23
USD
101884-640
101884-642
Methyl-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate ≥95%
Methyl-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₁NO₂S Melting Pt: 142…143 °C |
MDL Number:
MFCD11975799 CAS Number: 226259-30-9 |
Specification Test Results
Melting point | 142-143°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...