Methyl-2,6-dichloro-5-iodo-4-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2,6-dichloro-5-iodopyrimidine-4-carboxylate
048625-500MG
048625-1G
101884-634EA
903.08
USD
101884-634
101884-636
Methyl-2,6-dichloro-5-iodo-4-pyrimidinecarboxylate ≥95%
Methyl-2,6-dichloro-5-iodo-4-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₆H₃Cl₂IN₂O₂ MW: 332.91 g/mol Melting Pt: 92…93 °C |
MDL Number:
MFCD12922698 CAS Number: 1136962-00-9 |
Specification Test Results
Melting point | 92-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...