Methyl-6-bromo-4-hydroxyquinoline-8-carboxylate ≥95%
Supplier: Matrix Scientific
048623-500MG
048623-1G
101884-626EA
369.23
USD
101884-626
101884-628
Methyl-6-bromo-4-hydroxyquinoline-8-carboxylate ≥95%
Methyl 6-bromo-4-hydroxyquinoline-8-carboxylate
Hazard: Irritant
Formula:
C₁₁H₈BrNO₃ Melting Pt: 220-222°C |
MDL Number:
MFCD12922675 CAS Number: 1190198-14-1 |
Specification Test Results
Melting point | 220-222°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...