Methyl-5-bromo-4,6-dihydroxynicotinate ≥95%
Supplier: Matrix Scientific
048621-500MG
048621-1G
101884-620EA
261.54
USD
101884-620
101884-622
Methyl-5-bromo-4,6-dihydroxynicotinate ≥95%
Methyl-5-bromo-4,6-dihydroxynicotinate
Hazard: Irritant
Formula:
C₇H₆BrNO₄ MW: 248.04 g/mol Melting Pt: 254…256 °C (dec.) |
MDL Number:
MFCD12922713 CAS Number: 1190198-25-4 |
Specification Test Results
Melting point | 254-256(de |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...