Methyl-7-(benzyloxy)-1H-indole-5-carboxylate ≥95%
Supplier: Matrix Scientific
048620-500MG
048620-1G
101884-616EA
1089.23
USD
101884-616
101884-618
Methyl-7-(benzyloxy)-1H-indole-5-carboxylate ≥95%
Methyl-7-(benzyloxy)-1H-indole-5-carboxylate
Hazard: Irritant
Formula:
C₁₇H₁₅NO₃ Melting Pt: 145…147 °C |
MDL Number:
MFCD12922683 CAS Number: 1190198-13-0 |
Specification Test Results
Melting point | 145-147°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...