Methyl-3-amino-4-(methylamino)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-4-(methylamino)benzoate

048619-500MG 048619-1G
101884-612EA 115.38 USD
101884-612 101884-614
Methyl-3-amino-4-(methylamino)benzenecarboxylate ≥95%
Methyl-3-amino-4-(methylamino)benzenecarboxylate

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MW: 180.21 g/mol
Melting Pt: 103…105 °C
MDL Number: MFCD12922710
CAS Number: 66315-16-0

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR