Methyl-4-amino-3-(benzyloxy)-5-[2-(trimethylsilyl)ethynyl]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
048617-500MG
048617-1G
101884-604EA
747.69
USD
101884-604
101884-606
Methyl-4-amino-3-(benzyloxy)-5-[2-(trimethylsilyl)ethynyl]benzenecarboxylate ≥95%
Methyl-4-amino-3-(benzyloxy)-5-[2-(trimethylsilyl)ethynyl]benzenecarboxylate
Hazard: Irritant
Formula:
C₂₀H₂₃NO₃Si Melting Pt: 103…105 °C |
MDL Number:
MFCD12922682 CAS Number: 1190198-35-6 |
Specification Test Results
Melting point | 103-105°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...