2-[5-Methoxy-2-nitro-4-(trifluoromethyl)phenyl]acetic acid ≥95%
Supplier: Matrix Scientific
048615-500MG
048615-1G
101884-594EA
556.92
USD
101884-594
101884-596
2-[5-Methoxy-2-nitro-4-(trifluoromethyl)phenyl]acetic acid ≥95%
2-[5-Methoxy-2-nitro-4-(trifluoromethyl)phenyl]acetic acid
Hazard: Irritant
Formula:
C₁₀H₈F₃NO₅ Melting Pt: 191…193 °C |
MDL Number:
MFCD12922677 CAS Number: 1190198-33-4 |
Specification Test Results
Melting point | 191-193°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...