2-Iodo-4-nitro-5-(trifluoromethyl)aniline ≥95%

Supplier: Matrix Scientific

048614-500MG 048614-1G 048614-5G
101884-588EA 369.23 USD
101884-588 101884-590 101884-592
2-Iodo-4-nitro-5-(trifluoromethyl)aniline ≥95%
2-Iodo-4-nitro-5-(trifluoromethyl)aniline

Hazard: Irritant

Formula: C₇H₄F₃IN₂O₂
Melting Pt: 128-130°C
MDL Number: MFCD12922673
CAS Number: 852569-37-0

Order Now

Specification Test Results

Melting point 128-130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR