3-Iodo-2-methoxy-4-pyridinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-3-iodo-2-methoxypyridine

048613-500MG 048613-1G 048613-5G
101884-582EA 480 USD
101884-582 101884-584 101884-586
3-Iodo-2-methoxy-4-pyridinamine ≥95%
3-Iodo-2-methoxy-4-pyridinamine

Hazard: Irritant

Formula: C₆H₇IN₂O
MW: 250.04 g/mol
Melting Pt: 68…69 °C
MDL Number: MFCD12922699
CAS Number: 1190198-21-0

Order Now

Specification Test Results

Melting point 68-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR