4-Hydroxy-5-methoxy-2-pyridinecarbaldehyde ≥95%
Supplier: Matrix Scientific
048611-500MG
048611-1G
101884-576EA
921.54
USD
101884-576
101884-578
4-Hydroxy-5-methoxy-2-pyridinecarbaldehyde ≥95%
4-Hydroxy-5-methoxy-2-pyridinecarbaldehyde
Hazard: Irritant
Formula:
C₇H₇NO₃ Melting Pt: 173…174 °C |
MDL Number:
MFCD12922688 CAS Number: 204847-72-3 |
Specification Test Results
Melting point | 173-174°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...