5-Fluoro-1-benzothiophene-2-carbonyl chloride ≥95%
Supplier: Matrix Scientific
048608-500MG
048608-1G
101884-568EA
747.69
USD
101884-568
101884-570
5-Fluoro-1-benzothiophene-2-carbonyl chloride ≥95%
5-Fluoro-1-benzothiophene-2-carbonyl chloride
Hazard: Irritant
Formula:
C₉H₄ClFOS Melting Pt: 90…92 °C |
MDL Number:
MFCD12922700 CAS Number: 1190198-22-1 |
Specification Test Results
Melting point | 90-92°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...