Ethyl-4-hydroxy-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

048607-500MG 048607-1G
101884-564EA 375.38 USD
101884-564 101884-566
Ethyl-4-hydroxy-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate ≥95%
Ethyl-4-hydroxy-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₉H₁₁NO₄
Melting Pt: 117…119 °C
MDL Number: MFCD09031522
CAS Number: 853105-37-0

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR