Ethyl-5-bromo-4,6-dihydroxynicotinate ≥95%
Supplier: Matrix Scientific
048606-500MG
048606-1G
101884-560EA
369.23
USD
101884-560
101884-562
Ethyl-5-bromo-4,6-dihydroxynicotinate ≥95%
Ethyl-5-bromo-4,6-dihydroxynicotinate
Hazard: Irritant
Formula:
C₈H₈BrNO₄ Melting Pt: 267…269 °C |
MDL Number:
MFCD01411758 CAS Number: 89978-58-5 |
Specification Test Results
Melting point | 267-269°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...