Ethyl-1-benzyl-6-chloro-4-hydroxy-2-oxo-1,2-dihydro[1,8]naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

048605-500MG 048605-1G
101884-556EA 921.54 USD
101884-556 101884-558
Ethyl-1-benzyl-6-chloro-4-hydroxy-2-oxo-1,2-dihydro[1,8]naphthyridine-3-carboxylate ≥95%
Ethyl-1-benzyl-6-chloro-4-hydroxy-2-oxo-1,2-dihydro[1,8]naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₈H₁₅ClN₂O₄
Melting Pt: 152…154 °C
MDL Number: MFCD00946902
CAS Number: 179063-99-1

Order Now

Specification Test Results

Melting point 152-154°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR