Ethyl-2-(benzylamino)-5-iodonicotinate ≥95%

Supplier: Matrix Scientific

048603-500MG 048603-1G
101884-550EA 556.92 USD
101884-550 101884-552
Ethyl-2-(benzylamino)-5-iodonicotinate ≥95%
Ethyl-2-(benzylamino)-5-iodonicotinate

Hazard: Irritant

Formula: C₁₅H₁₅IN₂O₂
MW: 382.2 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD12922692
CAS Number: 1190198-19-6

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR