1-[6-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(6-Chloro-5-(trifluoromethyl)pyridin-2-yl)piperazine

048597-500MG 048597-1G
101884-526EA 556.92 USD
101884-526 101884-528
1-[6-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine ≥95%
1-[6-Chloro-5-(trifluoromethyl)-2-pyridinyl]piperazine

Hazard: Irritant

Formula: C₁₀H₁₁ClF₃N₃
Melting Pt: 76…78 °C
MDL Number: MFCD00892750
CAS Number: 132834-56-1

Order Now

Specification Test Results

Melting point 76-78°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR