4-(2-Chloro-3-pyridinyl)pyrimidine ≥95%

Supplier: Matrix Scientific

048596-500MG 048596-1G
101884-522EA 615.38 USD
101884-522 101884-524
4-(2-Chloro-3-pyridinyl)pyrimidine ≥95%
4-(2-Chloro-3-pyridinyl)pyrimidine

Hazard: Irritant

Formula: C₉H₆ClN₃
MW: 191.62 g/mol
Melting Pt: 85…86 °C
MDL Number: MFCD12922706
CAS Number: 870221-17-3

Order Now

Specification Test Results

Melting point 85-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR