4-(2-Chloro-3-pyridinyl)-N-methyl-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

048595-500MG 048595-1G
101884-518EA 746.15 USD
101884-518 101884-520
4-(2-Chloro-3-pyridinyl)-N-methyl-2-pyrimidinamine ≥95%
4-(2-Chloro-3-pyridinyl)-N-methyl-2-pyrimidinamine

Hazard: Irritant

Formula: C₁₀H₉ClN₄
Melting Pt: 187…189 °C
MDL Number: MFCD12922716
CAS Number: 870221-22-0

Order Now

Specification Test Results

Melting point 187-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR