1-(3-Chloro-5-nitro-1H-indazol-1-yl)-1-ethanone ≥95%

Supplier: Matrix Scientific

048594-500MG 048594-1G
101884-514EA 375.38 USD
101884-514 101884-516
1-(3-Chloro-5-nitro-1H-indazol-1-yl)-1-ethanone ≥95%
1-(3-Chloro-5-nitro-1H-indazol-1-yl)-1-ethanone

Hazard: Irritant

Formula: C₉H₆ClN₃O₃
MW: 239.62 g/mol
Melting Pt: 135…137 °C
MDL Number: MFCD00099555
CAS Number: 68159-07-9

Order Now

Specification Test Results

Melting point 135-137°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR