3-Amino-7-chloro-1H-indazole ≥97%
Supplier: Matrix Scientific
Synonyms:
7-Chloro-1H-indazol-3-amine
048588-500MG
048588-1G
048588-5G
101884-488EA
350.77
USD
101884-488
101884-490
101884-492
3-Amino-7-chloro-1H-indazole ≥97%
3-Amino-7-chloro-1H-indazole
Hazard: Irritant
Formula:
C₇H₆ClN₃ MW: 167.5956 g/mol Melting Pt: 187-189°C |
MDL Number:
MFCD07385618 CAS Number: 88805-67-8 |
Specification Test Results
Melting point | 187-189°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...