2-Chloro-6-(hydroxymethyl)-3-pyridinol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-(hydroxymethyl)pyridin-3-ol

048587-500MG 048587-1G 048587-5G
101884-482EA 369.23 USD
101884-482 101884-484 101884-486
2-Chloro-6-(hydroxymethyl)-3-pyridinol ≥95%
2-Chloro-6-(hydroxymethyl)-3-pyridinol

Hazard: Irritant

Formula: C₆H₆ClNO₂
MW: 159.57 g/mol
Melting Pt: 133…135 °C
MDL Number: MFCD12922671
CAS Number: 208519-41-9

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR