(5-Chloro-1-benzofuran-2-yl)(4-methoxyphenyl)methanone ≥95%
Supplier: Matrix Scientific
048585-500MG
048585-1G
048585-5G
101884-474EA
181.54
USD
101884-474
101884-476
101884-478
(5-Chloro-1-benzofuran-2-yl)(4-methoxyphenyl)methanone ≥95%
(5-Chloro-1-benzofuran-2-yl)(4-methoxyphenyl)methanone
Hazard: Irritant
Formula:
C₁₆H₁₁ClO₃ Melting Pt: 136…137 °C |
MDL Number:
MFCD12922712 CAS Number: 220526-76-1 |
Specification Test Results
Melting point | 136-137°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...