[2-Bromo-6-(trifluoromethyl)-3-pyridinyl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3-(hydroxymethyl)-6-(trifluoromethyl)pyridine

048584-500MG 048584-1G
101884-470EA 747.69 USD
101884-470 101884-472
[2-Bromo-6-(trifluoromethyl)-3-pyridinyl]methanol ≥95%
[2-Bromo-6-(trifluoromethyl)-3-pyridinyl]methanol

Hazard: Irritant

Formula: C₇H₅BrF₃NO
Melting Pt: 67…68 °C
MDL Number: MFCD11848029
CAS Number: 888738-18-9

Order Now

Specification Test Results

Melting point 67-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR