6-Bromo-8-nitro-4(1H)-quinolinone ≥95%

Supplier: Matrix Scientific

048582-500MG 048582-1G
101884-462EA 375.38 USD
101884-462 101884-464
6-Bromo-8-nitro-4(1H)-quinolinone ≥95%
6-Bromo-8-nitro-4(1H)-quinolinone

Hazard: Irritant

Formula: C₉H₅BrN₂O₃
Melting Pt: 259…261 °C
MDL Number: MFCD12922676
CAS Number: 1190198-29-8

Order Now

Specification Test Results

Melting point 259-261°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR