1-(5-Bromo-6-chloro-2-methyl-3-pyridinyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
048579-500MG
048579-1G
101884-452EA
892.31
USD
101884-452
101884-454
1-(5-Bromo-6-chloro-2-methyl-3-pyridinyl)-1-ethanone ≥95%
1-(5-Bromo-6-chloro-2-methyl-3-pyridinyl)-1-ethanone
Hazard: Irritant
Formula:
C₈H₇BrClNO Melting Pt: 47…49 °C |
MDL Number:
MFCD12922686 CAS Number: 1190198-15-2 |
Specification Test Results
Melting point | 47-49°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...