6-Bromo-1-benzothiophene-2-carbonyl chloride ≥95%
Supplier: Matrix Scientific
048576-500MG
048576-1G
101884-444EA
556.92
USD
101884-444
101884-446
6-Bromo-1-benzothiophene-2-carbonyl chloride ≥95%
6-Bromo-1-benzothiophene-2-carbonyl chloride
Hazard: Irritant
Formula:
C₉H₄BrClOS Melting Pt: 121…123 °C |
MDL Number:
MFCD12922705 CAS Number: 105212-27-9 |
Specification Test Results
Melting point | 121-123°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...