[2-(Benzyloxy)-4-methoxyphenyl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (2-(Benzyloxy)-4-methoxyphenyl)methanol

048575-500MG 048575-1G
101884-440EA 375.38 USD
101884-440 101884-442
[2-(Benzyloxy)-4-methoxyphenyl]methanol ≥95%
[2-(Benzyloxy)-4-methoxyphenyl]methanol

Hazard: Irritant

Formula: C₁₅H₁₆O₃
MW: 244.29 g/mol
Melting Pt: 68…69 °C
MDL Number: MFCD12922687
CAS Number: 171817-14-4

Order Now

Specification Test Results

Melting point 68-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR