5-(Benzyloxy)-2-(hydroxymethyl)-4-pyridinol ≥95%
Supplier: Matrix Scientific
048573-500MG
048573-1G
101884-436EA
375.38
USD
101884-436
101884-438
5-(Benzyloxy)-2-(hydroxymethyl)-4-pyridinol ≥95%
5-(Benzyloxy)-2-(hydroxymethyl)-4-pyridinol
Hazard: Irritant
Formula:
C₁₃H₁₃NO₃ Melting Pt: 228…230 °C |
MDL Number:
MFCD12922670 CAS Number: 165948-37-8 |
Specification Test Results
Melting point | 228-230°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...