5-(Benzyloxy)-2-(hydroxymethyl)-4H-pyran-4-one ≥95%

Supplier: Matrix Scientific

048572-500MG 048572-1G 048572-5G
101884-430EA 269.23 USD
101884-430 101884-432 101884-434
5-(Benzyloxy)-2-(hydroxymethyl)-4H-pyran-4-one ≥95%
5-(Benzyloxy)-2-(hydroxymethyl)-4H-pyran-4-one

Hazard: Irritant

Formula: C₁₃H₁₂O₄
Melting Pt: 132…134 °C
MDL Number: MFCD01927596
CAS Number: 15771-06-9

Order Now

Specification Test Results

Melting point 132-134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR