2-(Benzylamino)-5-iodonicotinic acid ≥95%
Supplier: Matrix Scientific
048570-500MG
048570-1G
101884-422EA
375.38
USD
101884-422
101884-424
2-(Benzylamino)-5-iodonicotinic acid ≥95%
2-(Benzylamino)-5-iodonicotinic acid
Hazard: Irritant
Formula:
C₁₃H₁₁IN₂O₂ Melting Pt: 210 °C (dec.) |
MDL Number:
MFCD12922691 CAS Number: 1190198-18-5 |
Specification Test Results
Melting point | 210(Dec)°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...