1H-1,2,3-Benzotriazol-4-amine ≥95%
Supplier: Matrix Scientific
048569-500MG
048569-1G
101884-418EA
556.92
USD
101884-418
101884-420
1H-1,2,3-Benzotriazol-4-amine ≥95%
1H-1,2,3-Benzotriazol-4-amine
Hazard: Irritant
Formula:
C₆H₆N₄ Melting Pt: 148…150 °C |
MDL Number:
MFCD00962622 CAS Number: 18076-61-4 |
Specification Test Results
Melting point | 148-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...