1-(2-Amino-5-iodo-3-methylphenyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
048567-500MG
048567-1G
048567-5G
101884-406EA
556.92
USD
101884-406
101884-408
101884-410
1-(2-Amino-5-iodo-3-methylphenyl)-1-ethanone ≥95%
1-(2-Amino-5-iodo-3-methylphenyl)-1-ethanone
Hazard: Irritant
Formula:
C₉H₁₀INO Melting Pt: 165…168 °C |
MDL Number:
MFCD11113358 CAS Number: 935292-72-1 |
Specification Test Results
Melting point | 165-168°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...