2-Amino-6-bromo-3-quinolinecarbonitrile ≥95%

Supplier: Matrix Scientific

048564-500MG 048564-1G
101884-392EA 461.54 USD
101884-392 101884-394
2-Amino-6-bromo-3-quinolinecarbonitrile ≥95%
2-Amino-6-bromo-3-quinolinecarbonitrile

Hazard: Irritant

Formula: C₁₀H₆BrN₃
Melting Pt: >300 °C
MDL Number: MFCD12922674
CAS Number: 951221-67-3

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR