7-Allyl-6-hydroxy-1-indanone ≥95%

Supplier: Matrix Scientific

048563-500MG 048563-1G
101884-388EA 556.92 USD
101884-388 101884-390
7-Allyl-6-hydroxy-1-indanone ≥95%
7-Allyl-6-hydroxy-1-indanone

Hazard: Irritant

Formula: C₁₂H₁₂O₂
Melting Pt: 123…125 °C
MDL Number: MFCD12922696
CAS Number: 320574-77-4

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR