1-(tert-Butyl)-4-(trifluoromethyl)-1H-pyrrolo-[2,3-b]pyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

048561-500MG 048561-1G
101884-380EA 375.38 USD
101884-380 101884-382
1-(tert-Butyl)-4-(trifluoromethyl)-1H-pyrrolo-[2,3-b]pyridine-3-carbonitrile ≥95%
1-(tert-Butyl)-4-(trifluoromethyl)-1H-pyrrolo-[2,3-b]pyridine-3-carbonitrile

Hazard: Irritant

Formula: C₁₃H₁₂F₃N₃
Melting Pt: 173…176 °C
MDL Number: MFCD12827805
CAS Number: 1146221-86-4

Order Now

Specification Test Results

Melting point 173-176°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR