7-(tert-Butyl)-2,4-bis(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine-5-carboxamide ≥95%

Supplier: Matrix Scientific

048558-500MG 048558-1G
101884-368EA 556.92 USD
101884-368 101884-370
7-(tert-Butyl)-2,4-bis(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine-5-carboxamide ≥95%
7-(tert-Butyl)-2,4-bis(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine-5-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₂F₆N₄O
Melting Pt: >140 °C (dec.)
MDL Number: MFCD12827804
CAS Number: 1186404-55-6

Order Now

Specification Test Results

Melting point >140Dec°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR